accessibility menu, dialog, popup

UserName

BFFFFH3CNCH3NN

CAS RN 59016-56-7

IUPAC Name: 1-cyano-4-(dimethylamino)pyridin-1-ium; tetrafluoroboranuide
Synonyms: 1-cyano-4-(dimethylamino)pyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 234.99
Molecular Formula: C8H10BF4N3
InChi Key: MBLVMDCQDCVKNE-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C1=CC=[N+](C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)