CAS RN 590-90-9
CAS RN 590-90-9
IUPAC Name:
4-hydroxybutan-2-one
Synonyms:
4-hydroxy-2-butanone
Molecular Weight (g/mol):
88.11
Molecular Formula:
C4H8O2
InChi Key:
LVSQXDHWDCMMRJ-UHFFFAOYSA-N
SMILES:
CC(=O)CCO
1
–
1
of
1
results
1
4-Hydroxy-2-butanone, 95%
CAS: 590-90-9 Molecular Formula: C4H8O2 Molecular Weight (g/mol): 88.106 MDL Number: MFCD00059005 InChI Key: LVSQXDHWDCMMRJ-UHFFFAOYSA-N Synonym: 4-hydroxy-2-butanone,2-butanone, 4-hydroxy,methylolacetone,3-oxo-1-butanol,3-oxobutanol,3-ketobutan-1-ol,monomethylolacetone,4-butanol-2-one,unii-tcm0bj44mf,2-hydroxyethyl methyl ketone PubChem CID: 111509 ChEBI: CHEBI:41268 IUPAC Name: 4-hydroxybutan-2-one SMILES: CC(=O)CCO