accessibility menu, dialog, popup

UserName

KOO

CAS RN 590-29-4

IUPAC Name: potassium formate
Synonyms: potassium formate hcook formic acid potassium salt potassium formiate formic acid, K salt
Molecular Weight (g/mol): 84.12
Molecular Formula: CHKO2
InChi Key: WFIZEGIEIOHZCP-UHFFFAOYSA-M
SMILES: [K+].[O-]C=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)