accessibility menu, dialog, popup

UserName

CAS RN 59-97-2

HClNNH

CAS RN 59-97-2

IUPAC Name: hydrogen 2-benzyl-4,5-dihydro-1H-imidazole chloride
Synonyms: hydrogen tolazoline chloride
Molecular Weight (g/mol): 196.68
Molecular Formula: C10H13ClN2
InChi Key: RHTNTTODYGNRSP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C(C1=NCCN1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)