accessibility menu, dialog, popup

UserName

CAS RN 589-92-4

CH3O

CAS RN 589-92-4

IUPAC Name: 4-methylcyclohexan-1-one
Synonyms: 4-methylcyclohexanone
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: VGVHNLRUAMRIEW-UHFFFAOYSA-N
SMILES: CC1CCC(=O)CC1
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)