CAS RN 589-82-2
CAS RN 589-82-2
IUPAC Name:
heptan-3-ol
Synonyms:
3-heptanol
Molecular Weight (g/mol):
116.2
Molecular Formula:
C7H16O
InChi Key:
RZKSECIXORKHQS-UHFFFAOYNA-N
SMILES:
CCCCC(O)CC
1
–
3
of
3
results
1
3-Heptanol, 98%
CAS: 589-82-2 Molecular Formula: C7H16O Molecular Weight (g/mol): 116.2 InChI Key: RZKSECIXORKHQS-UHFFFAOYSA-N Synonym: 3-heptanol,3-hydroxyheptane,ethyl butyl carbinol,butyl ethyl carbinol,butylethylcarbinol,xi-3-heptanol,fema no. 3547,ethylbutylcarbinol,heptanol-3,3-heptyl alcohol PubChem CID: 11520 IUPAC Name: heptan-3-ol SMILES: CCCCC(CC)O
2
(±)-3-Heptanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(±)-3-Heptanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.