accessibility menu, dialog, popup

UserName

OOCl

CAS RN 58880-37-8

IUPAC Name: 1-(4-chlorophenyl)cyclohexane-1-carboxylate
Synonyms: 1-(4-chlorophenyl)cyclohexane-1-carboxylate
Molecular Weight (g/mol): 237.7
Molecular Formula: C13H14ClO2
InChi Key: UPNXUJXIIZGXLQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results