accessibility menu, dialog, popup

UserName

NBrClNH2

CAS RN 588729-99-1

IUPAC Name: 5-bromo-2-chloropyridin-3-amine
Synonyms: 5-bromo-2-chloropyridin-3-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: ZSEZSALOLWCCGT-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results