accessibility menu, dialog, popup

UserName

SHClCl

CAS RN 5858-17-3

IUPAC Name: 3,4-dichlorobenzene-1-thiol
Synonyms: 3,4-dichlorobenzenethiol
Molecular Weight (g/mol): 179.06
Molecular Formula: C6H4Cl2S
InChi Key: HNJZDPKMMZXSKT-UHFFFAOYSA-N
SMILES: SC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)