accessibility menu, dialog, popup

UserName

H3CH3CCH3ClO

CAS RN 5856-77-9

IUPAC Name: 2,2-dimethylbutanoyl chloride
Synonyms: 2,2-dimethyl-butyryl chloride
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: LDJUYMIFFNTKOI-UHFFFAOYSA-N
SMILES: CCC(C)(C)C(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)