CAS RN 585566-00-3
IUPAC Name:
5-(2-fluorophenyl)furan-2-carbonyl chloride
Synonyms:
5-(2-fluorophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol):
224.62
Molecular Formula:
C11H6ClFO2
InChi Key:
VKIMJQXHHBMFER-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1C1=CC=C(O1)C(Cl)=O
1
–
1
of
1
results
1
5-(2-Fluorophenyl)-2-furoyl chloride, Thermo Scientific™
CAS: 585566-00-3 Molecular Formula: C11H6ClFO2 Molecular Weight (g/mol): 224.615 MDL Number: MFCD02258009 InChI Key: VKIMJQXHHBMFER-UHFFFAOYSA-N Synonym: 5-2-fluorophenyl furan-2-carbonyl chloride,5-2-fluorophenyl-2-furoyl chloride,acmc-20am9v,5-2-fluorophenyl-2-furancarbonyl chloride,5-2-fluoro-phenyl-furan-2-carbonyl chloride PubChem CID: 3530559 IUPAC Name: 5-(2-fluorophenyl)furan-2-carbonyl chloride SMILES: C1=CC=C(C(=C1)C2=CC=C(O2)C(=O)Cl)F