accessibility menu, dialog, popup

UserName

BrBr

CAS RN 58556-75-5

IUPAC Name: 2,7-dibromonaphthalene
Synonyms: 2,7-dibromonaphthalene
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: ODJZWBLNJKNOJK-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC(Br)=CC=C2C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results