accessibility menu, dialog, popup

UserName

CAS RN 58530-53-3

IUPAC Name: 2,4-dibromopyridine
Synonyms: 2,4-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: PCMMSLVJMKQWMQ-UHFFFAOYSA-N
SMILES: BrC1=CC=NC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results