accessibility menu, dialog, popup

UserName

CAS RN 58442-17-4

OHNN

CAS RN 58442-17-4

IUPAC Name: 1H-1,3-benzodiazole-6-carbaldehyde
Synonyms: 3H-1,3-benzodiazole-5-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: ALCHVVTYAHQOFY-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2N=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results