accessibility menu, dialog, popup

UserName

CAS RN 584-84-9

CH3NCONCO

CAS RN 584-84-9

IUPAC Name: 2,4-diisocyanato-1-methylbenzene
Synonyms: toluene diisocyanate TDI 2,4/2,6-toluenediisocyanate 2,4-tolylene diisocyanate tolylene-2,4-diisocyanate toluene diisocyanate (mixture of isomers)
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: DVKJHBMWWAPEIU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N=C=O)N=C=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)