accessibility menu, dialog, popup

UserName

NH2NNN

CAS RN 584-13-4

IUPAC Name: 4H-1,2,4-triazol-4-amine
Synonyms: 4H-1,2,4-triazole, 4-amino-
Molecular Weight (g/mol): 84.08
Molecular Formula: C2H4N4
InChi Key: FMCUPJKTGNBGEC-UHFFFAOYSA-N
SMILES: NN1C=NN=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)