CAS RN 5836-29-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Coumatetralyl, TRC
CAS: 5836-29-3 Molecular Formula: C19 H16 O3 Molecular Weight (g/mol): 292.33 IUPAC Name: 4-hydroxy-3-(1,2,3,4-tetrahydronaphthalen-1-yl)chromen-2-one SMILES: OC1=C(C2CCCc3ccccc23)C(=O)Oc4ccccc14
2
Coumatetralyl, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Coumatetralyl, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.