accessibility menu, dialog, popup

UserName

CAS RN 583-59-5

CH3HO

CAS RN 583-59-5

IUPAC Name: 2-methylcyclohexan-1-ol
Synonyms: 2-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: NDVWOBYBJYUSMF-UHFFFAOYNA-N
SMILES: CC1CCCCC1O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)