accessibility menu, dialog, popup

UserName

CAS RN 583-05-1

H3COO

CAS RN 583-05-1

IUPAC Name: 1-phenylpentane-1,4-dione
Synonyms: 1-phenylpentane-1,4-dione
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: RBLXWIPBPPVLPU-UHFFFAOYSA-N
SMILES: CC(=O)CCC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results