accessibility menu, dialog, popup

UserName

CAS RN 582-52-5

IUPAC Name: 5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Synonyms: 5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: KEJGAYKWRDILTF-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)