accessibility menu, dialog, popup

UserName

CAS RN 58161-35-6

CH3ONHO

CAS RN 58161-35-6

IUPAC Name: N-(1-oxo-2,3-dihydro-1H-inden-5-yl)acetamide
Synonyms: N-(1-oxo-2,3-dihydroinden-5-yl)acetamide
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: GHUPGGYDRVSZSW-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C2C(=O)CCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results