accessibility menu, dialog, popup

UserName

OHOHSOOOSOOOH2OH2ONaNa

CAS RN 5808-22-0

IUPAC Name: 4,5-dihydroxynaphthalene-2,7-disulfonate
Synonyms: 4,5-dihydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 318.27
Molecular Formula: C10H6O8S2
InChi Key: HLVXFWDLRHCZEI-UHFFFAOYSA-L
SMILES: OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (13)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (9)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (8)
  • (3)
  • (3)
  • (1)
  • (13)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)