CAS RN 58069-82-2
CAS RN 58069-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Urea-13C, TRC
CAS: 58069-82-2 Molecular Formula: 13C H4 N2 O Molecular Weight (g/mol): 61.05 Synonym: Urea 13C IUPAC Name: diamino(1^{13}C)methanone SMILES: N[13C](=O)N