accessibility menu, dialog, popup

UserName

CH3NH2CH3NH2H3CH2N

CAS RN 58052-80-5

IUPAC Name: 2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
Synonyms: acetaldehyde ammonia trimer
Molecular Weight (g/mol): 132.23
Molecular Formula: C6H18N3
InChi Key: MZSSRMMSFLVKPK-UHFFFAOYSA-Q
SMILES: CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (4)
Boiling Point 
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (3)