accessibility menu, dialog, popup

UserName

NOOSOOCl

CAS RN 58032-84-1

IUPAC Name: (3-nitrophenyl)methanesulfonyl chloride
Synonyms: (3-nitrophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 235.64
Molecular Formula: C7H6ClNO4S
InChi Key: IUYFKLDRPZLAQQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(CS(Cl)(=O)=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results