accessibility menu, dialog, popup

UserName

HOOHOHONOHNO

CAS RN 58-96-8

IUPAC Name: 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: β-uridine allo-uridine allo uridine allouridine uridine (substance) arauridine 1-β-delta-ribofuranosyluracil
Molecular Weight (g/mol): 244.2
Molecular Formula: C9H12N2O6
InChi Key: DRTQHJPVMGBUCF-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
Molecular Weight (g/mol) 
  • (9)
  • (9)
Quantity 
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (6)
Percent Purity 
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (9)
  • (9)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (6)
  • (7)
  • (2)