CAS RN 58-72-0
IUPAC Name:
(2,2-diphenylethenyl)benzene
Synonyms:
(2,2-diphenylethenyl)benzene benzilidenediphenylmethane
Molecular Weight (g/mol):
256.35
Molecular Formula:
C20H16
InChi Key:
MKYQPGPNVYRMHI-UHFFFAOYSA-N
SMILES:
C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenylethylene, 98+%
CAS: 58-72-0 Molecular Formula: C20H16 Molecular Weight (g/mol): 256.348 MDL Number: MFCD00004765 InChI Key: MKYQPGPNVYRMHI-UHFFFAOYSA-N Synonym: triphenylethylene,triphenylethene,ethylene, triphenyl,benzilidenediphenylmethane,1,1,2-triphenylethylene,ethene-1,1,2-triyltribenzene,1,2-diphenylvinyl benzene,1,2,2-triphenylethylene,benzene, 1,1',1-1-ethenyl-2-ylidene tris,unii-s4zlz1k74b PubChem CID: 6025 ChEBI: CHEBI:35034 IUPAC Name: 1,2-diphenylethenylbenzene SMILES: C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3