accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 5798-75-4

IUPAC Name: ethyl 4-bromobenzoate
Synonyms: ethyl 4-bromobenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: XZIAFENWXIQIKR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results