accessibility menu, dialog, popup

UserName

CAS RN 5794-28-5

H2OCaOOOO

CAS RN 5794-28-5

IUPAC Name: calcium hydrate oxalate
Synonyms: calcium hydrate oxalate
Molecular Weight (g/mol): 146.11
Molecular Formula: C2H2CaO5
InChi Key: LQHWSGSWNOHVHO-UHFFFAOYSA-L
SMILES: O.[Ca++].[O-]C(=O)C([O-])=O
Physical Form 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)