accessibility menu, dialog, popup

UserName

OOHNH2ONH2H2O

CAS RN 5794-24-1

IUPAC Name: 2-amino-3-carbamoylpropanoic acid hydrate
Synonyms: asparagin hydrate
Molecular Weight (g/mol): 150.13
Molecular Formula: C4H10N2O4
InChi Key: RBMGJIZCEWRQES-UHFFFAOYNA-N
SMILES: O.NC(CC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (6)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)