accessibility menu, dialog, popup

UserName

H2ONH2(S)H2NOOHO

CAS RN 5794-13-8

IUPAC Name: (2S)-2-amino-3-carbamoylpropanoic acid hydrate
Synonyms: L-asparagine hydrate
Molecular Weight (g/mol): 150.13
Molecular Formula: C4H10N2O4
InChi Key: RBMGJIZCEWRQES-DKWTVANSSA-N
SMILES: O.N[C@@H](CC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (13)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (7)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)
  • (13)
  • (1)
  • (4)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)