accessibility menu, dialog, popup

UserName

CH3OOHOO(S)(R)OO(R)(R)CH3(R)O(R)CH3O(R)OHCH3H2C

CAS RN 57444-62-9

IUPAC Name: [(1R,2R,6R,10S,11R,15R,17R)-13-benzyl-6-hydroxy-4,17-dimethyl-5-oxo-15-(prop-1-en-2-yl)-12,14,18-trioxapentacyclo[11.4.1.0¹,¹⁰.0²,⁶.0¹¹,¹⁵]octadeca-3,8-dien-8-yl]methyl 2-(4-hydroxy-2-methoxyphenyl)acetate
Synonyms: resiniferatoxin reciniferatoxin RTX
Molecular Weight (g/mol): 628.72
Molecular Formula: C37H40O9
InChi Key: OYWHJFQVSOUQSF-PKRQWDGKSA-N
SMILES: COC1=CC(O)=CC=C1CC(=O)OCC1=C[C@H]2[C@H]3OC4(CC5=CC=CC=C5)O[C@]3(C[C@@H](C)[C@]2(O4)[C@@H]2C=C(C)C(=O)[C@@]2(O)C1)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results