accessibility menu, dialog, popup

UserName

CAS RN 5743-47-5

H2OH2OH2OH2OH2OCaH3COHOOH3COHOO

CAS RN 5743-47-5

IUPAC Name: calcium bis(2-hydroxypropanoate) pentahydrate
Synonyms: calcium pentahydrate dilactate
Molecular Weight (g/mol): 308.29
Molecular Formula: C6H20CaO11
InChi Key: HPVJXNNKHRNBOY-UHFFFAOYNA-L
SMILES: O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results