accessibility menu, dialog, popup

UserName

OHOBrNOO

CAS RN 573-54-6

IUPAC Name: 2-bromo-3-nitrobenzoic acid
Synonyms: 2-bromo-3-nitrobenzoic acid
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: WTDJEGSXLFHZPY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1Br)[N+]([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results