accessibility menu, dialog, popup

UserName

CAS RN 57297-29-7

HClH2NNH

CAS RN 57297-29-7

IUPAC Name: hydrogen cyclopropanecarboximidamide chloride
Synonyms: hydrogen cyclopropanecarboximidamide chloride
Molecular Weight (g/mol): 120.58
Molecular Formula: C4H9ClN2
InChi Key: JRYOZJIRAVZGMV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results