CAS RN 5728-33-6
IUPAC Name:
3'-methyl-[1,1'-biphenyl]-4-carboxylic acid
Synonyms:
3'-methyl-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol):
212.25
Molecular Formula:
C14H12O2
InChi Key:
CVFLWXCNPWFJEH-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=C1)C1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
3'-Methylbiphenyl-4-carboxylic acid, 96%, Thermo Scientific™
CAS: 5728-33-6 Molecular Formula: C14H12O2 Molecular Weight (g/mol): 212.25 MDL Number: MFCD03424590 InChI Key: CVFLWXCNPWFJEH-UHFFFAOYSA-N Synonym: 3'-methylbiphenyl-4-carboxylic acid,4-3-methylphenyl benzoic acid,3'-methyl-1,1'-biphenyl-4-carboxylic acid,3'-methyl-biphenyl-4-carboxylic acid,3'-methyl biphenyl-4-carboxylic acid,3'-methyl-4-biphenylcarboxylic acid,rarechem al be 1342,pubchem10295,acmc-1apyt,4-m-tolyl-benzoic acid PubChem CID: 2759816 IUPAC Name: 4-(3-methylphenyl)benzoic acid SMILES: CC1=CC=CC(=C1)C1=CC=C(C=C1)C(O)=O