accessibility menu, dialog, popup

UserName

CAS RN 57196-62-0

ClCH3ONH2

CAS RN 57196-62-0

IUPAC Name: 6-methoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Synonyms: 6-methoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Molecular Weight (g/mol): 199.68
Molecular Formula: C10H14ClNO
InChi Key: QIUIKPQXMJJOQT-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC=C2C[NH2+]CCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results