accessibility menu, dialog, popup

UserName

CAS RN 57102-42-8

IUPAC Name: 9-(4-bromophenyl)-9H-carbazole
Synonyms: 9-(4-bromophenyl)carbazole
Molecular Weight (g/mol): 322.21
Molecular Formula: C18H12BrN
InChi Key: XSDKKRKTDZMKCH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results