accessibility menu, dialog, popup

UserName

CH3CH3

CAS RN 571-61-9

IUPAC Name: 1,5-dimethylnaphthalene
Synonyms: 1,5-dimethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: SDDBCEWUYXVGCQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=C(C)C=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results