accessibility menu, dialog, popup

UserName

CAS RN 570-23-0

NH2OHOHO

CAS RN 570-23-0

IUPAC Name: 3-amino-2-hydroxybenzoic acid
Synonyms: 3-aminosalicylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: IQGMRVWUTCYCST-UHFFFAOYSA-N
SMILES: NC1=CC=CC(C(O)=O)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)