accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 57-97-6

IUPAC Name: 7,12-dimethyltetraphene
Synonyms: 7,12-dimethylbenz(a)anthracene 7,12-dime-benz(A)anthracene 7,12-dimethylbenzanthracene 7,12-dimethylbenzanthrancene 9,10-dimethylbenz(a)anthracene
Molecular Weight (g/mol): 256.35
Molecular Formula: C20H16
InChi Key: ARSRBNBHOADGJU-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=CC=CC=C3C2=C(C)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)