accessibility menu, dialog, popup

UserName

CAS RN 57-57-8

IUPAC Name: oxetan-2-one
Synonyms: β propiolactone 3-propiolactone caswell no. 709 β-propanoic acid lactone β-propionolactone BPL β-proprolactone betaprone propiolattone β-propiolactone
Molecular Weight (g/mol): 72.06
Molecular Formula: C3H4O2
InChi Key: VEZXCJBBBCKRPI-UHFFFAOYSA-N
SMILES: O=C1CCO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results