accessibility menu, dialog, popup

UserName

CAS RN 57-09-0

IUPAC Name: hexadecyltrimethylazanium bromide
Synonyms: cetyltrimethylammonium bromide centimide ctmab cee dee cetrimonii bromidum bromuro de cetrimonio cetab cetyl trimethyl ammonium bromide bromure de cetrimonium cetrimonium bromide
Molecular Weight (g/mol): 364.46
Molecular Formula: C19H42BrN
InChi Key: LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (4)
  • (23)
Quantity 
  • (2)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
Percent Purity 
  • (6)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (5)
  • (4)
Grade 
  • (1)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (4)
  • (23)
  • (2)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (5)
  • (4)
  • (1)
  • (4)
  • (1)