accessibility menu, dialog, popup

UserName

ClBrOH

CAS RN 56962-04-0

IUPAC Name: 3-bromo-5-chlorophenol
Synonyms: 3-bromo-5-chlorophenol
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: GMGWXLPFRHYWAS-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results