accessibility menu, dialog, popup

UserName

HClNH2H2NNH

CAS RN 569-61-9

IUPAC Name: hydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Synonyms: pararosaniline
Molecular Weight (g/mol): 323.82
Molecular Formula: C19H18ClN3
InChi Key: JUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (3)
Quantity 
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
Grade 
  • (7)
Physical Form 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results