accessibility menu, dialog, popup

UserName

CH3SNNOClHCl

CAS RN 568577-86-6

IUPAC Name: 1-benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonyl chloride hydrochloride
Synonyms: 3-benzyl-2-(methylsulfanyl)imidazole-4-carbonyl chloride hydrochloride
Molecular Weight (g/mol): 303.2
Molecular Formula: C12H12Cl2N2OS
InChi Key: AUZZRVMDIAHPNZ-UHFFFAOYSA-N
SMILES: Cl.CSC1=NC=C(N1CC1=CC=CC=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results