accessibility menu, dialog, popup

UserName

CAS RN 568577-82-2

CH3ONCO

CAS RN 568577-82-2

IUPAC Name: 3-isocyanato-2-methyl-5-phenylfuran
Synonyms: 3-isocyanato-2-methyl-5-phenylfuran
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: LVNIEPBRJLDJHU-UHFFFAOYSA-N
SMILES: CC1=C(C=C(O1)C1=CC=CC=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results