CAS RN 56812-60-3
IUPAC Name:
magnesium(2+) (2-bromophenyl)methanide bromide
Synonyms:
magnesium(2+) (2-bromophenyl)methanide bromide
Molecular Weight (g/mol):
274.24
Molecular Formula:
C7H6Br2Mg
InChi Key:
XGAWDDBPRRNVDO-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].[CH2-]C1=CC=CC=C1Br