CAS RN 5673-07-4
IUPAC Name:
1,3-dimethoxy-2-methylbenzene
Synonyms:
1,3-dimethoxy-2-methylbenzene
Molecular Weight (g/mol):
152.19
Molecular Formula:
C9H12O2
InChi Key:
FPEUDBGJAVKAEE-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(OC)=C1C
1
–
4
of
4
results
1
2,6-Dimethoxytoluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Dimethoxytoluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,6-Dimethoxytoluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2,6-Dimethoxytoluene, 98%
CAS: 5673-07-4