CAS RN 56704-25-7
IUPAC Name:
1-(2,4,6-trimethylpyridin-3-yl)ethan-1-one
Synonyms:
1-(2,4,6-trimethylpyridin-3-yl)ethanone
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
RKNKTACHKMTWTD-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(C)C=C(C)N=C1C
1
–
1
of
1
results
1
3-Acetyl-2,4,6-trimethylpyridine, 98%, Thermo Scientific™
CAS: 56704-25-7 Molecular Formula: C10H13NO Molecular Weight (g/mol): 163.22 MDL Number: MFCD00085730 InChI Key: RKNKTACHKMTWTD-UHFFFAOYSA-N Synonym: 3-acetyl-2,4,6-trimethylpyridine,1-2,4,6-trimethylpyridin-3-yl ethanone,1-2,4,6-trimethylpyridin-3-yl ethan-1-one,1-2,4,6-trimethyl-3-pyridinyl ethanone,ethanone,1-2,4,6-trimethyl-3-pyridinyl,3-acetyl-collidine,acmc-20anas,pubchem9575,ethanone, 1-2,4,6-trimethyl-3-pyridinyl-9ci PubChem CID: 268124 IUPAC Name: 1-(2,4,6-trimethylpyridin-3-yl)ethanone SMILES: CC1=CC(=NC(=C1C(=O)C)C)C